Egg-Shaped Spinbar™ Magnetic Stir Bars
Egg-Shaped Spinbar™ Magnetic Stir Bars
Actual product may vary
Bel-Art
Egg-Shaped Spinbar™ Magnetic Stir Bars
Designed for use in round-bottom vessels
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Diameter (Metric)Length (English)Diameter (English)
FIS22-26173450.8 mm19 mm2 in.0.75 in.
FIS07-910-2316 mm6 mm0.625 in.0.23 in.
FIS22-1271009.5 mm4.7 mm0.37 in.0.18 in.
Catalog number FIS22-261734
Price (SGD)
95.00
EA
Add to cart
Length (Metric):
50.8 mm
Diameter (Metric):
19 mm
Length (English):
2 in.
Diameter (English):
0.75 in.
Price (SGD)
95.00
EA
Add to cart
  • Provide their own pivot point without a separate ring or abrupt change in contour for unrestricted spin
  • Minimal contact when used in plastic containers
  • Magnet itself is egg-shaped encapsulated in FDA grade, USP Class VI Teflon™ PTFE, except three smallest sizes have a cylindrical magnet in egg-shaped Teflon PTFE
  • Manufactured in a registered ISO 9001:2008, as verified by SGS certification
  • Autoclave safe
Specifications
AutoclavableAutoclavable
Certifications/ComplianceFDA, USP Class VI, ISO 9001:2008, SGS
ColorWhite
Diameter (English)0.75 in.
For Use With (Equipment)2000 and 3000mL round-bottom flasks
Length (English)2 in.
CoatingPTFE, FDA-grade, USP Class VI
Diameter (Metric)19 mm
Length (Metric)50.8 mm
MaterialAlnico Magnet
Quantity1
ShapeEgg
TypeMagnetic Stir Bar
Unit SizeEA