Long-Handled Dippers
Long-Handled Dippers
Actual product may vary
Bel-Art
Long-Handled Dippers
Welded handles provide easy sampling from tanks, vats, lakes, etc. Floatable, high-density polyethylene plastic dippers will not rust or corrode and provide chemical resistance.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Length (Metric) HandleLength (English) HandleIncludes
FIS22-2615701000 mL1.8 m6 ft.One-Piece Handle
FIS22-111153500 mL0.9 m3 ft.One-Piece Handle
FIS22-163790500 mL3.7 m12 ft.Two-Piece Handle
Catalog number FIS22-261570
Price (SGD)
338.00
EA
Add to cart
Capacity (Metric):
1000 mL
Length (Metric) Handle:
1.8 m
Length (English) Handle:
6 ft.
Includes:
One-Piece Handle
Price (SGD)
338.00
EA
Add to cart
Welded handles provide easy sampling from tanks, vats, lakes, etc. Floatable, high-density polyethylene plastic dippers will not rust or corrode and provide chemical resistance.
  • High-density polyethylene dippers will not rust or corrode
  • Provides excellent chemical resistance
  • Handle is welded to the rigid bowl at a 45° angle
  • End of the handle has a hook for hanging
  • Handle diameter: 29 mm (1.13 in.)
Specifications
Product TypeDipper
MaterialHDPE
AutoclavableNot Autoclavable
IncludesOne-Piece Handle
Handle TypeLong
Capacity (English)32 oz.
Capacity (Metric)1000 mL
For Use With (Application)For easy sampling from tanks, vats, and lakes
Diameter (English) Handle1.13 in.
Diameter (Metric) Handle2.9 cm
Length (English) Handle6 ft.
Length (Metric) Handle1.8 m
Unit SizeEA