Rotary Separatory Funnel Racks
Actual product may vary
Rotary Separatory Funnel Racks
Rotary Separatory Funnel Racks
Rotary Separatory Funnel Racks
Bel-Art
Rotary Separatory Funnel Racks
Consists of a rectangular base, screw-in column and a support disc held in place by a screw clamp
Have Questions?
Catalog NumberPositions
FIS22-2606686
Catalog number FIS22-260668
Price (SGD)
799.00
Each
Positions:
6
Price (SGD)
799.00
Each
  • The support disc is fully adjustable up to 21 in.
  • Support holes for funnels are open front type for easy removal and placement of separatory funnels
  • Includes 18 insert clips used to accommodate different sized separatory funnels with a capacity of 500 mL to 1000 mL
Specifications
AutoclavableNot Autoclavable
ColorWhite
HoldsFunnel (500 to 1000 mL)
MaterialSteel
Positions6
SterilityNon-sterile
For Use With (Equipment)Seperatory Funnels
Size350 x 530 mm
Dimensions (L x W x H)21 x 21 x 21 in. (53.34 x 53.34 x 53cm)
Length (Metric)53.34 cm
Length (English)21 in.
Width (English)21 in.
Height (English)21 in.
IncludesBase, Uprights, Racks, Insert Clips
Product TypeFunnel Rack
Height (Metric)53 cm
Width (Metric)53.34 cm
Unit SizeEach