Polypropylene Water Bath Rack
Polypropylene Water Bath Rack
Actual product may vary
Bel-Art
Polypropylene Water Bath Rack
Submersible, sinks without adding weight
Have Questions?
Catalog NumberDimensions (L x W x H)For Use With (Equipment)Holds
FIS22-26062221.3 x 11 x 11.1 cm13 to 16 mm Test Tubes50 Test Tubes
Catalog number FIS22-260622
Price (SGD)
165.00
EA
Add to cart
Dimensions (L x W x H):
21.3 x 11 x 11.1 cm
For Use With (Equipment):
13 to 16 mm Test Tubes
Holds:
50 Test Tubes
Price (SGD)
165.00
EA
Add to cart
  • Durable polypropylene rack that sinks to the bottom of a water bath and remains upright when agitated
  • Steam autoclavable at 121°C (250°F)
  • Dimensions: 8.375L x 4.312 in.W (213 x 110mm)
Specifications
AutoclavableAutoclavable
DescriptionWater Bath Tube Rack
For Use With (Equipment)13 to 16 mm Test Tubes
TypeTest Tube Rack
Holds50 Test Tubes
MaterialPolypropylene
Dimensions (L x W x H)21.3 x 11 x 11.1 cm
Length (English)8.37 in.
Length (Metric)21.3 cm
Width (English)4.31 in.
Width (Metric)11 cm
Height (English)4.37 in.
Height (Metric)11.1 cm
Diameter (Metric) Well16 mm
Unit SizeEA