Poxygrid™ Drain Stand
Actual product may vary
Poxygrid™ Drain Stand
Poxygrid™ Drain Stand
Bel-Art
Poxygrid™ Drain Stand
Features three rows of epoxy-coated hairpin-loop supports (14 in all) to hold labware with 18 mm I.D. necks or larger
Have Questions?
Catalog NumberPositions
FIS22-26059426
Catalog number FIS22-260594
Price (SGD)
479.00
Each
Positions:
26
Price (SGD)
479.00
Each
Features three rows of epoxy-coated hairpin-loop supports (14 in all) to hold labware with 18 mm I.D. necks or larger
  • Top row has 12 straight pins 10.2 cm (4 in.) long to accommodate smaller items
  • A polystyrene tray catches water as labware drains
  • Rubber tipped feet provide non-skid use on a tabletop, or the stand may be wall-mounted using wire loops on the back
Specifications
AutoclavableNot Autoclavable
ColorBlue
HoldsLab Glassware
MaterialPolystyrene
Positions26
ShapeTriangle
StackableNo
SterilityNon-sterile
For Use With (Equipment)Lab Glassware
Size430 x 170 x 480 mm
Length (Metric)17 cm
Length (English)6.5 in.
Width (English)16.75 in.
Height (English)19 in.
IncludesDrip Tray
Product TypeDraining Rack
ShelvesNone
Height (Metric)48 cm
Array12 x 1, 5 x 2, 4 x 1
Width (Metric)43 cm
Unit SizeEach