Poxygrid™ Wire Test Tube Racks
Poxygrid™ Wire Test Tube Racks
Actual product may vary
Poxygrid™ Wire Test Tube Racks
Poxygrid™ Wire Test Tube Racks
Bel-Art
Poxygrid™ Wire Test Tube Racks
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorPositions
FIS22-260553Blue72
FIS11-391-311Blue108
Catalog number FIS22-260553
Price (SGD)
164.00
Each
Add to cart
Color:
Blue
Positions:
72
Price (SGD)
164.00
Each
Add to cart
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
  • Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
  • Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
  • Orange racks can be used for biohazard warning while yellow for radioactive warning
  • Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
  • Stores from 40 to 108 tubes (five different diameters of testtubes)
  • Autoclavable
Specifications
AutoclavableAutoclavable
ColorBlue
HoldsTubes (13 to 16 mm)
MaterialEpoxy-coated Steel
Positions72
ShapeRectangular
StackableYes, When Empty
SterilityNon-sterile
Dimensions (L x W x H)9.5 x 5.1 x 2.5 in. (242 x 130 x 64 mm)
Length (Metric)242 mm
Length (English)9.48 in.
Width (English)5.12 in.
Height (English)2.51 in.
Diameter (Metric) Opening16 mm
Product TypeTest Tube Rack
Shelves3 Tiers
Height (Metric)64 mm
Array6 x 12
Width (Metric)130 mm
Unit SizeEach