Polypropylene Powder Funnels
Polypropylene Powder Funnels
Actual product may vary
Bel-Art
Polypropylene Powder Funnels
Feature a large bore stem to minimize bridging of powder
Have Questions?
Catalog NumberCapacity (Metric)Diameter (Metric) Stem
FIS22-260284784 mL2.9 cm
Catalog number FIS22-260284
Price (SGD)
92.00
Pack
Add to cart
Capacity (Metric):
784 mL
Diameter (Metric) Stem:
2.9 cm
Price (SGD)
92.00
Pack
Add to cart
  • 60° angle
  • Steam autoclavable at 121°C (250°F)
  • Stem length 25 mm
Specifications
Capacity (Metric)784 mL
Diameter (Metric) Stem2.9 cm
MaterialPolypropylene
Height (Metric)13.6 cm
Angle60°
AutoclavableAutoclavable
For Use With (Application)For transferring powders
Length (Metric) Stem2.5 cm
Product TypeFilling Funnel (Powders)
Unit SizePack