HDPE Dispensing Jug
HDPE Dispensing Jug
Actual product may vary
Bel-Art
HDPE Dispensing Jug
Used to store and dispense common solutions, including janitorial cleaners, deionized and distilled water, or other aqueous liquids.
Have Questions?
Catalog NumberCapacity (Metric)MaterialProduct Type
FIS22-26010820 LHDPEDispensing Jug
Catalog number FIS22-260108
Price (SGD)
185.00
EA
Add to cart
Capacity (Metric):
20 L
Material:
HDPE
Product Type:
Dispensing Jug
Price (SGD)
185.00
EA
Add to cart
  • Storage capacity is 20 liters (5 gallons)
  • Gradations in gallons are molded into the side
  • Jug made from strong and lightweight HDPE
  • Dimensions: 25 x 38 cm (9-7/8 x 15 in.)
Specifications
AutoclavableNot Autoclavable
Capacity (English)5.3 gal.
Capacity (Metric)20 L
Closure MaterialLDPE
ColorWhite
For Use With (Application)To store and dispense common solutions, including janitorial cleaners, deionized / distilled water, or other aqueous liquids
MaterialHDPE
Mouth StyleWide
Product TypeDispensing Jug