Book Bottle
Book Bottle
Actual product may vary
Bel-Art
Book Bottle
Polypropylene bottles stack book-like on shelves for efficient use of space.
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-2600685000 mL
Catalog number FIS22-260068
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
5000 mL
  • Features two handles: a top handle plus a convenient grip hold in the front facilitates removal from a shelf
  • Graduated markings on front in liters
Specifications
Capacity169.07 oz., 5000 mL
Capacity (Metric)5000 mL
ColorTranslucent, White
MaterialPolypropylene
Quantity1 Ea.
SterilityNon-sterile
Closure IncludedYes
AutoclavableAutoclavable
DisposableReusable
GraduatedYes
MouthWide, Threaded
ShapeSquare with Handle
Height (Metric)241.3 mm
Product TypeRectangular Bottle
Width (English)14.75 in.
Height (English)9.5 in.
Closure MaterialPolypropylene
Closure Size43 mm
Closure TypeThreaded
Unit SizeEA