Polystop™ Dropper Bottles
Polystop™ Dropper Bottles
Actual product may vary
Bel-Art
Polystop™ Dropper Bottles
Prevent chipping of the glass dropper upon reinsertion into the dropper bottle.
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-259515100 mL
Catalog number FIS22-259515
Price (SGD)
-
Capacity (Metric):
100 mL
  • The dropper easily pulls free, while the stopper remains snugly in the neck of the bottle, thus providing a safe return space for the dropper
  • Polypropylene stopper
  • Clear glass bottle has a ground T-neck, and the dropper has a soft vinyl bulb
  • Disassembles for easy cleaning
Specifications
Capacity3.4 oz., 100 mL
Capacity (Metric)100 mL
Capacity (English)3.4 oz.
ColorClear
MaterialGlass
Quantity1 Ea.
SterilityNon-sterile
Closure IncludedYes
AutoclavableNot Autoclavable
DisposableReusable
Dropper Capacity100 mL
Drop Size0.002 oz.
GraduatedNo
MouthNarrow, Threaded
ShapeCylindrical
Height (Metric)104.4 mm
Product TypeDropping Bottle
Width (English)2.1 in.
Height (English)4.11 in.
Closure MaterialPolypropylene
Diameter (Metric)53.3 mm
Closure TypeThreaded
Unit SizeEA