LabTemp™ 20 Surface Temperature Indicators
LabTemp™ 20 Surface Temperature Indicators
Actual product may vary
LabTemp™ 20 Surface Temperature Indicators
Accurate and continuous monitoring of temperatures between 1° to 21°C
Have Questions?
Catalog NumberTemperature Range (Metric)
FIS22-2461971°C to 21°C
Catalog number FIS22-246197
Price (SGD)
-
Temperature Range (Metric):
1°C to 21°C
  • Can be used with temperature-sensitive materials in laboratories specifically when these materials are taken in or out of a refrigerated environment
  • Designed for use in clinical chemistry, microbiology and laboratory areas where specific temperatures are required to keep compounds or organisms
  • Disposable and easily affix to plastic, glass, metal, and wood labware — compatible with all types of surfaces for up to 18 months
  • Can be used on items stored in refrigerators, temperature controlled centrifuges, low temperature incubators, and even water baths for 1-2 hours
Specifications
For Use With (Application)Monitoring refrigerators and freezers, instant and continuous temperature
Product TypeSurface Temperature Indicator
Max. Temperature (Metric)21°C
Min. Temperature (Metric)1°C
Temperature Range (Metric)1°C to 21°C
Unit SizeCS