HemoTemp™ II Reversible/Irreversible Blood Temperature Indicator
Actual product may vary
HemoTemp™ II Reversible/Irreversible Blood Temperature Indicator
Reversible and irreversible indicator
Have Questions?
Catalog Number
TemperatureRangeMetric
FIS22-246194
Catalog number FIS22-246194
Price (SGD)
339.00
PK
Add to cart
Price (SGD)
339.00
PK
Add to cart
Reversible indicator acts as a blood bag thermometer calibrated to display the internal temperature of the bag under ambient conditions
The irreversible portion signals that the internal blood bag temperature has reached or exceeded 10°C; the signal continues even if the blood is recooled
Should not be the sole method for evaluating the clinical acceptability of blood
Specifications
For Use With (Application)Time and temperature history of the blood unit