Check Order Status
Y Connectors
Actual product may vary
Bel-Art
Y Connectors
Use these fittings to ensure a tight, secure connection between lengths of tubing having the same inner diameter (I.D.)
Have Questions?
Catalog Number
For Use With
FIS22-235531
6mm
Catalog number
FIS22-235531
Price (SGD)
150.00
PK
Add to cart
For Use With:
6mm
Price (SGD)
150.00
PK
Add to cart
Multiple barbs per arm provide additional contact between the fitting and the tubing for extra gripping power
Chemical-resistant polypropylene plastic for use with a wide range of liquids or gases
FDA-grade for direct contact with food, drugs, and cosmetics
Autoclavable
Specifications
Autoclavable
Autoclavable
Material
Polypropylene
For Use With
6mm
Shape
Y
Color
Natural
Product Type
Y Tubing Connector