Pipet Filler
Pipet Filler
Actual product may vary
Bel-Art
Pipet Filler
Squeeze, then release the bulb to draw fluid into the pipet
Have Questions?
Catalog number FIS22-235309
Price (SGD)
145.00
PK
Add to cart
Price (SGD)
145.00
PK
Add to cart
Squeeze, then release the bulb to draw fluid into the pipet
  • The pipet aid is removed when fluid fills the bulb, and the pipet top quickly covered with a gloved finger to safely and easily control and release fluid
  • The bulb is Vikem™ vinyl with a tapered polyethylene chuck for quick removal when pipet is filled
Specifications
AutoclavableNot Autoclavable
ColorBlue
DisposableReusable
For Use WithPipette
Grip TypeRound
MaterialVinyl
ModelStandard
Number of Channels1
SterilityNon-sterile
TypePipette Filler
For Use With (Equipment)Pipet
Unit SizePK