Surgicutt™ Bleeding Time Device
Actual product may vary
Accriva Diagnostics
Surgicutt™ Bleeding Time Device
Standardized incision depth avoids deep cuts and ensures an incision well above concentrated pain fibers
Have Questions?
Catalog NumberProduct Type
FIS22-211-506Bleeding Time Device
Catalog number FIS22-211-506
Price (SGD)
-
Product Type:
Bleeding Time Device
  • Unique, Surgicutt technology provides a uniform incision, not a puncture, improving test standardization and reproducibility of bleeding time test results
  • Stainless-steel blade is a permanent retractable blade—safe and avoids disease transmission
  • Controlled depth and length of incision every time: large blade slot surface reduces skin indentation
  • Virtually painless—diminished bruising means less trauma and more comfort for patients
  • Color coded, disposable, and easy to use
  • Available in three sizes: adult, pediatric, and infant
Specifications
Blade MaterialStainless Steel
Depth (English)0.039 in.
Depth (Metric)0.1cm
DisposableSingle-use
For Use With (Application)Avoids deep cuts and ensures an incision well above concentrated pain fibers, used to screen for platelet function
Lancet StyleRetractable blade
Length (English)0.19 in.
Length (Metric)0.5cm
SizeAdult
Product TypeBleeding Time Device
Unit SizePack