Pasteur Pipets
Pasteur Pipets
Actual product may vary
Pasteur Pipets
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialTypeLength (English)Quantity
FIS22-209379Borosilicate GlassCotton-plugged Pasteur Pipet5.75 in.Case of 4 x 250
FIS22-063156Soda Lime GlassEasel5.75 in.200/Cs.
FIS22-042816Soda Lime GlassEasel5.75 in.250/Cs.
FIS22-209353Soda Lime GlassCotton-plugged Pasteur Pipet5.75 in.Case of 4 x 250
FIS22-209361Soda Lime GlassEasel9 in.Case of 4 x 250
FIS22-063172Soda Lime GlassEasel9 in.200/Cs.
FIS22-183624Borosilicate GlassPasteur Pipet5.75 in.4 packs of 250 in one box
FIS22-183632Borosilicate GlassEasel9 in.1000 Case
Catalog number FIS22-209379
Price (SGD)
411.00
CS
Add to cart
Material:
Borosilicate Glass
Type:
Cotton-plugged Pasteur Pipet
Length (English):
5.75 in.
Quantity:
Case of 4 x 250
Price (SGD)
411.00
CS
Add to cart
Being packaged in strong corrugated material provides maximum protection from external shock for Fisherbrand™ Pasteur Pipets. Each pack also contains a foam pad to protect the pipet tips.
  • Straight-cut tips and smooth, fire-polished tops to facilitate bulb attachment
  • Positive constriction approximately 1 in. from the top to prevent cotton plugs from slipping into pipet barrel
  • Easel configuration of each pack keeps pipets orderly and easily accessible
Specifications
Diameter (English) Outer0.27 in.
Diameter (Metric) Outer7 mm
MaterialBorosilicate Glass
Length (English)5.75 in.
QuantityCase of 4 x 250
Tip StyleStraight
Top StyleSmooth fire polished
Volume (Metric)2 mL
Length (Metric)14.6 cm
TypeCotton-plugged Pasteur Pipet
Unit SizeCS