HYSYNAL™ Synthetic Rubber Disposable Tourniquet Straps
Actual product may vary
HYSYNAL™ Synthetic Rubber Disposable Tourniquet Straps
HYSYNAL™ Synthetic Rubber Disposable Tourniquet Straps
HYSYNAL™ Synthetic Rubber Disposable Tourniquet Straps
HYSYNAL™ Synthetic Rubber Disposable Tourniquet Straps
HYSYNAL™ Synthetic Rubber Disposable Tourniquet Straps
Hygenic
HYSYNAL™ Synthetic Rubber Disposable Tourniquet Straps
Contain no latex proteins and provide a safe alternative for latex-sensitive patients
Have Questions?
Catalog NumberWidth (Metric)
FIS22-203-5625.1 mm
Catalog number FIS22-203-562
Price (SGD)
963.00
Case
Width (Metric):
5.1 mm
Price (SGD)
963.00
Case
Straps are constructed of a special formulation of synthetic rubber that provides a nonslip powder-free surface while still performing like natural latex. You can easily tie and untie these tear-resistant straps.
  • Designed for disposable use in hospitals and clinics
  • Nonlatex for patients with latex sensitivities; latex protein free
  • Variety of packaging options are available
  • Individually precut to measure 1W x 18 in.L (2.5 x 45.7cm)
  • Tear-resistant and slip-resistant
  • Easily tied and untied; can be rolled and banded
  • Thickness: 0.6mm
  • Color: light blue
  • Perforated every 18 in. for easy dispensing
Specifications
ColorLight Blue
Latex FreeYes
MaterialRubber
Product TypeTourniquet Strap
For Use With (Application)Designed for disposable use in hospitals and clinics
DisposableSingle-use
Thickness (English)0.02 in.
Thickness (Metric)0.6 mm
Width (Metric)5.1 mm
Unit SizeCase