Biohazard Sharp Object Safety Pouch Stand
Biohazard Sharp Object Safety Pouch Stand
Actual product may vary
Bel-Art
Biohazard Sharp Object Safety Pouch Stand
Offers protection against cuts by contaminated sharp objects during work in labs, clinics, and medical offices
Have Questions?
Catalog number FIS22-197061
View Price:Sign in to see your account pricing. Need an account? Register with us today.
  • Provides a stable support to keep the Biohazard Sharp Object Safety Pouch ( Mfr. No. 13234-0000) upright and open for easy access
  • Constructed from bright orange, epoxy-coated steel wire
  • Frame dimensions are 10 x 13 x 20 cm H (4 x 5.125 x 8 in.)
Specifications
Capacity (English)1 safety pouch
ColorWhite
AutoclavableNot Autoclavable
For Use With (Application)For providing stable support to biohazard sharp objects safety pouch
MaterialEpoxy-coated Steel Wire
Product TypeSharps Container
Unit SizeEA