Flanged Plug Caps For 13mm Tubes
Actual product may vary
Globe Scientific Inc
Flanged Plug Caps For 13mm Tubes
Provide leak-resistance in test tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS22-171-676Yellow
FIS22-171-668Blue
FIS22-171-675White
FIS22-171-672Lavender
FIS22-171-671Black
FIS22-171-670Green
FIS22-171-674Red
FIS22-171-673Orange
Catalog number FIS22-171-676
Price (SGD)
57.00
Pack
Color:
Yellow
Price (SGD)
57.00
Pack
  • Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
  • Available in nine colors for sample identification purposes
Specifications
Includes2 flexible flanges
Product TypeFlange Plug Cap
MaterialLDPE
ColorYellow
Size (English)0.51 in.
Size (Metric)13 mm
For Use With (Equipment)13 mm Tubes
Unit SizePack