Flanged Plug Caps For 12mm Tubes
Actual product may vary
Globe Scientific Inc
Flanged Plug Caps For 12mm Tubes
Provide leak-resistance in test tubes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
FIS22-171-664Black
FIS22-171-665Lavender
FIS22-171-666Orange
Catalog number FIS22-171-664
Price (SGD)
60.00
Pack
Color:
Black
Price (SGD)
60.00
Pack
  • Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
  • Available in nine colors for sample identification purposes
Specifications
Includes2 flexible flanges
Product TypeFlange Plug Cap
MaterialPolyethylene
ColorBlack
Size (English)0.47 in.
Size (Metric)12 mm
For Use With (Equipment)12 mm Tubes
Unit SizePack