Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Tubes
▸
Tube Caps and Closures
▸
Flanged Plug Caps For 16mm Tubes
Actual product may vary
Globe Scientific Inc
Flanged Plug Caps For 16mm Tubes
Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes
Have Questions?
Catalog Number
Color
FIS22-171-652
Orange
Catalog number
FIS22-171-652
Price (SGD)
88.00
Pack
Add to cart
Color:
Orange
Price (SGD)
88.00
Pack
Add to cart
Provide leak-resistance in test tubes, centrifuge tubes and round cuvettes
Polyethylene caps with two flexible flanges that press firmly against inside wall of tubes
Available in seven colors for sample identification purposes
Specifications
Includes
2 flexible flanges
Product Type
Flange Plug Cap
Material
Polyethylene
Color
Orange
Size (English)
0.62 in.
Size (Metric)
16 mm
For Use With (Equipment)
16 mm Tubes