12mL Centrifuge Tubes
Actual product may vary
Globe Scientific Inc
12mL Centrifuge Tubes
Polystyrene and Polypropylene tubes with conical bottoms and rim
Have Questions?
Catalog NumberMaterialClosure Included
FIS22-171-622PolystyreneNone
Catalog number FIS22-171-622
Price (SGD)
155.00
Case
Material:
Polystyrene
Closure Included:
None
Price (SGD)
155.00
Case
  • Available with graduations at 0.5, 1, 2.5 and 10mL
  • Centrifuge spin rate for Polystyrene tubes is 1500rcf and for Polypropylene tubes is 3000rcf
  • Dimensions: 16 x 100mm
Specifications
Diameter (English) Outside0.6 in.
Capacity (Metric)12 mL
MaterialPolystyrene
For Use WithBayer™ analyzers for urinalysis procedures to make pouring off easy
No. per Case2000
No. per Pack500
SterilityNon-sterile
ShapeConical
Length (Metric)100 mm
Capacity (English)0.405 oz.
Bottom ShapeConical
Closure TypeNot Included
Closure IncludedNone
Diameter (Metric)16 mm
GraduatedYes
Graduations0.5, 1, 2, 5 and 10 mL
Product TypeCentrifuge Tube
Max. RCF1,500 x g
Unit SizeCase