Polypropylene Transport Tubes
Actual product may vary
Globe Scientific Inc
Polypropylene Transport Tubes
Designed for safe storage and transport of biological materials
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-170-1935 mL
Catalog number FIS22-170-193
Price (SGD)
344.00
Case
Capacity (Metric):
5 mL
Price (SGD)
344.00
Case
  • Polypropylene allows unobstructed viewing of sample
  • Shatter-proof and may be exposed to freezing temperatures
  • Feature leak-resistant polyethylene screw caps that can be removed or sealed in single turn
  • Tube is self-standing with conical bottom and with molded graduations
Specifications
Closure ColorWhite
AutoclavableAutoclavable
Capacity (Metric)5 mL
Bottom ShapeConical
MaterialPolypropylene
Thread StyleExternal
IncludesWhite screw cap
No. per Case1000
SterilityNon-sterile
GraduatedYes
Closure MaterialPolyethylene
Height (Metric)56 mm
Closure TypeScrew Cap
Diameter (Metric)16 mm
Product TypeTransport Tube
Unit SizeCase