SP Scienceware™ B.O.D. Bottle Carrier
SP Scienceware™ B.O.D. Bottle Carrier
Actual product may vary
Bel-Art
SP Scienceware™ B.O.D. Bottle Carrier
Tall handle for easy carrying
Have Questions?
Catalog number FIS22-163782
Price (SGD)
-
Polypropylene carrier holds eight standard 300mL Biological Oxygen Demand bottles
  • Dimensions: 33 x 17 x 7cmH (13.125 x 6.25 x 2.625 in.); 9cm (3.625 in.)
  • Openings are 7.2cm diameter (2.8 in.)
  • Steam autoclavable at 121°C (250°F)
Specifications
Product TypeBottle Carrier
AutoclavableAutoclavable
HoldsBiological Oxygen Demand Bottles (300 mL)
MaterialPolypropylene
Positions8
Dimensions (L x W x H)6.75 x 13.12 x 2.62 in. (17 x 33 x 7cm)
Length (Metric)17 cm
Length (English)6.75 in.
Width (Metric)33 cm
Width (English)13.12 in.
Height (Metric)7 cm
Height (English)2.62 in.
Unit SizePK