DuoClick Specimen Containers
Actual product may vary
Fisherbrand
DuoClick Specimen Containers
User friendly, cost effective and safe alternative for specimen collection and transport
Have Questions?
Catalog NumberCapacity (Metric)Sterility
FIS22-150-24090 mLSterile
Catalog number FIS22-150-240
Price (SGD)
273.00
Case
Capacity (Metric):
90 mL
Sterility:
Sterile
Price (SGD)
273.00
Case
  • A simple turn of the closure will give user assurance that the seal is complete with a “click sound” and the second “click” ensures 95 kPa compliance and maximum leak-resistance for both pneumatic systems and transportation by air
  • Closure also features a visual cue, with an arrow that will align with the closure area once secure
  • Wide mouth design (53mm) is very convenient for patient use
  • Containers are graduated to aid in accurate measurement of patient specimen
  • Made in the U.S.A. using FDA compliant and prime virgin medical grade resins (closure is HDPE and container is Polypropylene)
  • Available in three convenient sizes (60mL, 90mL and 120mL) both sterile and non-sterile options
Specifications
Product TypeSpecimen Container
Closure IncludedYes
GraduatedYes
MaterialPolypropylene
SterilitySterile
Capacity (English)3 oz.
Capacity (Metric)90 mL
Certifications/Compliance95kPa compliance
Closure MaterialHDPE
Closure Size53 mm
Closure TypeDouble-click Screw Cap
For Use With (Application)Specimen collection and transport
Unit SizeCase