Drugs of Abuse Replenishment Supplies
Actual product may vary
Therapak
Drugs of Abuse Replenishment Supplies
Supplies for use with single- and split-specimen collection kits.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
FIS22-131-561Specimen Transport Bag
FIS22-131-560Tamper Evident Security Seal
Catalog number FIS22-131-561
Price (SGD)
129.00
Pack
Product Type:
Specimen Transport Bag
Price (SGD)
129.00
Pack
Specifications
Product TypeSpecimen Transport Bag
MaterialPolyethylene
SterilityNon-sterile
Closure TypePermanent Adhesive
For Use With (Application)Provides protection of specimen and sensitive patient information during the transport process
For Use With (Equipment)Single and split specimen collection kits in drugs of abuse testing
IncludesRequisition pouch, biohazard symbol and 3 x 4 in. absorbent sheet, removable adhesive release liner
Quantity100 Pk.
Volume (English)1.7 oz.
Volume (Metric)50 mL
Unit SizePack