Specifications
Product TypeTamper Evident Security Seal
MaterialAcetate Film with Paper Backing
SterilityNon-sterile
ColorRed
For Use With (Application)Used for chain of custody procedures in drug testing, identify the presence of any tampering with the specimen containers or vials from the moment they are sealed until they are opened at the testing laboratory
For Use With (Equipment)Single and split specimen collection kits in drugs of abuse testing
Quantity500 Pk.
Unit SizePack