Evidence Tape
Actual product may vary
Therapak
Evidence Tape
Perforated, self-adhesive security seal for maintaining chain of custody in drug testing procedures.
Have Questions?
Catalog NumberColor
FIS22-131-470Red
Catalog number FIS22-131-470
Price (SGD)
45.00
Case
Color:
Red
Price (SGD)
45.00
Case
Therapak™ Tamper Evident Perforated Security Seals are designed for chain of custody drug testing, featuring strategically positioned perforations for easy removal.
  • Material: Bright white paper
  • Color: Red
  • Dimensions: 0.75 in. x 6.625 in.
  • Identification Fields: Includes spaces for Donor Initials, Collector Signature, I.D. or SS#, and Date
  • Features: Tamper evident, easy-peel, self-adhesive design for strong adhesion to vials and jars
Specifications
Adhesive TypePermanent
ColorRed
MaterialAcetate Film with Paper Backing
Product TypeTamper-evident Label
For Use With (Application)Used for chain of custody procedures in drug testing
Quantity100/Cs.
Size (English)0.75 x 6.62 in.
Size (Metric)1.9 x 16.8 cm
Length (English)6.625 in.
Width (English)0.75 in.
Unit SizeCase