Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Mailing and Shipping Products
▸
Mailing and Shipping Containers
▸
Biological Substance Category B Ambient Shipping Systems
Actual product may vary
Therapak
Biological Substance Category B Ambient Shipping Systems
Include all the necessary marking labels in a convenient format for application at the shipping location
Have Questions?
Catalog Number
Capacity
Includes
FIS22-130-419
Holds 6 tubes
95kPa Bag with Tube Shuttle
Catalog number
FIS22-130-419
Price (SGD)
519.00
Case
Add to cart
Capacity:
Holds 6 tubes
Includes:
95kPa Bag with Tube Shuttle
Price (SGD)
519.00
Case
Add to cart
For transport of blood, urine and other biological substances
Choice of pre-assembled systems to accommodate a variety of shipping needs
Specifications
Quantity
1 Ea.
Includes
95kPa Bag with Tube Shuttle
Applications
For blood, urine transport
Type
Ambient Shipping for Biological Substance Category B
Capacity
Holds 6 tubes
Packaging
Transport box, 95kPa
Unit Size
Case
Contents & Storage
95kPa Bag; TearTech tube shuttle, cushioning material, biohazard and Biological Substance Category B labels, list of contents card, transport box