95kPa Compliant Transport Bags
Actual product may vary
Therapak
95kPa Compliant Transport Bags
For use as a secondary water-tight receptacle
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeIncludes
FIS22-130-4111-pocket Transport BagBag only
FIS22-130-0202-pocket Transport BagBag only
FIS22-130-0232-pocket Transport BagAqui-Pak Pouch
Catalog number FIS22-130-411
Price (SGD)
4,441.00
Case
Product Type:
1-pocket Transport Bag
Includes:
Bag only
Price (SGD)
4,441.00
Case
For use as a secondary water-tight receptacle
  • Leak-resistant seal
  • Back pocket holds quarter-folded requisition forms
  • To transport multiple primary tubes, use the Aqui-Pak™ absorbent and tube separator
  • Easy-to-use — simply insert specimen tube and absorbent material into the front pocket, peel away the white tape liner, and fold down adhesive to cover the opening
  • For transport of multiple primary tubes, use the Aqui-Pak absorbent and tube separator with a 95 kPa bag
  • For transport of cryogenic cryoboxes in a frozen shipper, use the 12 × 15 in. (30.5 × 38.1cm), one-pocket bag, which accommodates up to 3 in. (7.6cm) tall cryoboxes
Specifications
DescriptionOne-pocket bag
ColorClear with White Tape Liner
IncludesBag only
MaterialPlastic
Dimensions (L x W)15 x 12 in. (38.1 x 30.5cm)
Length (English)15 in.
Width (English)12 in.
Product Type1-pocket Transport Bag
Length (Metric)38.1 cm
Width (Metric)30.5 cm
Unit SizeCase