EPS (Expanded Polystyrene) Insulated Shippers
Actual product may vary
Therapak
EPS (Expanded Polystyrene) Insulated Shippers
For refrigerated and frozen specimen shipments
Have Questions?
Catalog NumberDimensions (L x W x H) ExteriorSizeDimensions (D x W x H) Interior
FIS22-130-40711 x 9.5 x 10.62 in.Small7.87 x 6.37 x 7.75 in.
Catalog number FIS22-130-407
Price (SGD)
314.00
Case
Dimensions (L x W x H) Exterior:
11 x 9.5 x 10.62 in.
Size:
Small
Dimensions (D x W x H) Interior:
7.87 x 6.37 x 7.75 in.
Price (SGD)
314.00
Case
  • Ideal insulated shippers for reference laboratories, pharmaceutical companies, hospitals and clinical testing laboratories who ship and receive temperature sensitive specimens
  • Wall thickness insulation ranges from 1 to 2 in. depending on cooler
  •  
    Extra Small Insulated Shipper
    • Holds up to two cold packs for a small refrigerated specimen shipment
  • Small Insulated Shipper
    • For routine specimen shipments of cold or frozen specimens in clinical trials studies
    • Accommodates one standard 2 or 3 in. cryostorage box
    • Primarily used as an overnight shipping system for a single patient set of specimen tubes
  • Medium Insulated Shipper
    • Holds up to two standard 2 or 3 in. cryostorage boxes
    • Ideal for shipping up to five small specimen bags of sample tubes for clinical trial site shipments to a laboratory
    Large Insulated Shipper
    • Holds up to 25 pounds of dry ice making it suitable for applications that involve frozen transport up to 72 hours
    • Used for overnight shipments of multiple patient specimen packages where dry ice capacity may be reduced to accommodate more specimen bags or crystorage boxes
Specifications
ApplicationsFor reference laboratories, pharmaceutical companies, hospitals and clinical testing laboratories
SizeSmall
Dimensions (L x W x H) Exterior11 x 9.5 x 10.62 in.
Dimensions (D x W x H) Interior7.87 x 6.37 x 7.75 in.
MaterialExpanded Polystyrene
Unit SizeCase