Gel Pack Protective Barrier Transport Bag
Actual product may vary
Therapak
Gel Pack Protective Barrier Transport Bag
Insulate specimen samples requiring ambient to cool environment during transit
Have Questions?
Catalog NumberDimensions (L x W)
FIS22-130-1109 x 6 in. (22.9 x 15.2cm)
Catalog number FIS22-130-110
Price (SGD)
399.00
Case
Dimensions (L x W):
9 x 6 in. (22.9 x 15.2cm)
Price (SGD)
399.00
Case
Just add cold water to designated line to form gel.
  • Lightweight, compact alternative to standard insulating coolants
  • 6 × 9 in. (15.2 × 22.9cm) zip style bag
  • Pillow pack materials are nontoxic and noncarcinogenic
  • Bags store flat and are virtually weightless until activated
Specifications
Dimensions (L x W)9 x 6 in. (22.9 x 15.2cm)
Length (English)9 in.
Width (English)6 in.
Length (Metric)22.9 cm
Width (Metric)15.2 cm
Unit SizeCase