Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See all Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Mailing and Shipping Products
▸
Mailing and Shipping Containers
▸
TearTech™ Tube Shuttles
Actual product may vary
Therapak
TearTech™ Tube Shuttles
Separates tubes to prevent contact
Have Questions?
Change view
Catalog Number
Closure Type
Quantity
FIS22-130-038
Zipper
Up to 8 tubes
FIS22-130-019
TearTech
Up to 6 tubes
2 Options
Catalog number
FIS22-130-038
Price (SGD)
1,065.00
Case
Add to cart
Closure Type:
Zipper
Quantity:
Up to 8 tubes
Price (SGD)
1,065.00
Case
Add to cart
Leak-tight construction for increased safety during transport
Numbered tube slots for easy reference
Two sizes available to hold six or eight blood or urine tubes (12–16mm dia.)
Six-tube shuttle features a TearTech closure that folds down for a permanent seal, yet provides a natural perforation for easy opening at the lab
Eight tube shuttle features a zip-style closure
Shuttles can be rolled or folded and placed inside a secondary watertight container
Specifications
Quantity
Up to 8 tubes
Applications
Blood and urine tube mailings
Closure Type
Zipper