For transport of blood tubes and urine vials and other biological substances
Have Questions?
Change view
Catalog Number
Quantity
Size
FIS22-130-037
Up to 20 tubes
Large
FIS22-130-035
Up to 5 tubes
Small
FIS22-130-036
Up to 12 tubes
Medium
3 Options
Catalog number FIS22-130-037
Price (SGD)
422.00
Case
Quantity:
Up to 20 tubes
Size:
Large
Price (SGD)
422.00
Case
Reusable containers with durable, thick-wall construction to protect specimens
Ideal secondary, watertight container
Screw cap for secure, watertight seal
Small canister constructed of translucent PET to identify specimen leakage; holds up to five tubes
Medium canister for harsh transport conditions including dry ice shipments of frozen specimens; holds up to 6 tubes
Large canister is most durable of canisters and can be used for biological substances transport between client sites and reference labs; holds up to 20 tubes
Specifications
QuantityUp to 20 tubes
Certifications/ComplianceComply with DOT and IATA regulations as a secondary container
ApplicationsFor transport of biological substances