Slide Tray Cabinet
Slide Tray Cabinet
Actual product may vary
Bel-Art
Slide Tray Cabinet
Compact cabinet to organize and store microscope slides
Have Questions?
Catalog number FIS22-088799
Price (SGD)
1,292.00
Each
Add to cart
Price (SGD)
1,292.00
Each
Add to cart
  • Small footprint cabinet can be placed on the benchtop, making slides easily accessible
  • Dust-free storage
  • Holds 240 slides in 12 microscope trays (Mfr. No. H44581-0000, not included)
  • Polypropylene outer shell; polystyrene tray rails
  • Transparent vertically sliding acrylic door with door knob
  • 35.6L x 20.8W x 25.4H cm (14 x 83/16 x 10 in.)
Specifications
TypeSlide Storage Cabinet
Slide Capacity240
Depth (English)14 in.
Depth (Metric)356 mm
Device Holds240
For Use With (Equipment)Microscope
Height (English)10 in.
Height (Metric)254 mm
MaterialPolypropylene, Polystyrene
Width (English)5.18 in.
Width (Metric)131 mm
Unit SizeEach