Microscope Slide Tray
Microscope Slide Tray
Actual product may vary
Bel-Art
Microscope Slide Tray
White polystyrene background provides good visual contrast for viewing dyes and stains
Have Questions?
Catalog number FIS22-088740
Price (SGD)
350.00
Pack
Add to cart
Price (SGD)
350.00
Pack
Add to cart
White polystyrene background provides good visual contrast for viewing dyes and stains
  • Stores slides in a horizontal position for full visibility
  • Handy finger cut-out simplifies removal of slides
  • Each tray stores 20 standard 1 x 3 in. (25 x 75mm) slides
  • Can be used independently or conveniently stored in the Slide Tray Cabinet (Mfr. No. 445830000, sold separately)
  • Dimensions: 6.5 x 13.5 in. (17 x 34cm)
Specifications
TypeSlide Storage Tray
Slide Capacity20
ColorWhite
Device Holds20
For Use With (Equipment)Microscope Slide