Cytoseal™ Mountant
Actual product may vary
Thermo Scientific
Cytoseal™ Mountant
Prevent cracking, fading and discoloration of stained specimens with Cytoseal™ mounting media.
Have Questions?
Catalog NumberQuantityProduct Line
FIS22-050-2624 oz.Cytoseal XYL
Catalog number FIS22-050-262
Price (SGD)
-
Quantity:
4 oz.
Product Line:
Cytoseal XYL
  • Cytoseal 60 mountant is a low-viscosity medium that dries quickly and allows for rapid, even spreading which virtually eliminates air bubbles
  • Cytoseal 280 mountant is a high-viscosity medium for applications where minimal spreading is desired or applications without a cover glass
  • Cytoseal XYL mountant is a xylene-based rapid drying medium that allows for microscopic examination of slides soon after application
Ordering Information:
Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.
Specifications
Packaging Type4 oz Dispenser Bottle
Product LineCytoseal XYL
Product TypeMountant
Quantity4 oz.
FormulationFast-drying formula reduces waiting time and increases throughput
For Use With (Application)Provides exceptional clarity and longevity for your microscopy slides
Unit SizeEach