Tissue Floatation Baths and Flotation Work Stations
Actual product may vary
Tissue Floatation Baths and Flotation Work Stations
Tissue Floatation Baths and Flotation Work Stations
Fisherbrand
Tissue Floatation Baths and Flotation Work Stations
Offer a choice of 8 x 8 x 2.25 in. or 8 x 11 x 2 in. work surfaces
Have Questions?
Catalog NumberAmperageDimensions (L x W x H)
FIS22-047-7733.5 A38.1 x 47.6 x 13.3 cm
Catalog number FIS22-047-773
Price (SGD)
-
Amperage:
3.5 A
Dimensions (L x W x H):
38.1 x 47.6 x 13.3 cm
  • Microprocessor-controlled heating from ambient to 65°C
  • High contrast background illuminated by a bright LED array provides optimal specimen viewing
  • Removable glass dish permits easy cleaning
  • Scratch-, heat- and chemical-resistant plastic housing
  • Magnetic on/off switch addresses reliability issues of competitive mechanical switches
Additional Features of Floatation Work Stations
  • 2 × 8 in. convection slide dryer
  • 2 × 2 in. HISTO/Orientator eliminates wrinkles
Specifications
Controller TypeMicroprocessor
Certifications/ComplianceUL, CSA, CE
TypeWater Bath
DisplayLED
Dimensions (L x W x H)38.1 x 47.6 x 13.3 cm
Length (English)15 in.
Length (Metric)38.1 cm
Width (English)18.75 in.
Width (Metric)47.6 cm
Height (English)5.25 in.
Height (Metric)13.3 cm
Weight (English)11 lb.
Weight (Metric)6 kg
Electrical Requirements100/120 V 50/60 Hz
Voltage100/120 V
Amperage3.5 A