ThermoSafe Nylon Carriers
ThermoSafe Nylon Carriers
Actual product may vary
ThermoSafe Nylon Carriers
Lightweight, nylon clad, durable units ideal for on-site collection, storage and transportation of temperature-sensitive specimens, medications, blood products, medical devices, and organs
Have Questions?
Catalog number FIS22-046788
Price (SGD)
284.00
EA
Add to cart
Price (SGD)
284.00
EA
Add to cart
Model 458
  • Washable, weather-resistant nylon shell with thick-walled EPS foam insulation
  • Adjustable nylon carrying straps for hand or shoulder carrying
  • Includes U-tek refrigerant
  • Capacity: 1.0 cu. ft.
Model 459
  • Washable, weather-resistant nylon shell with thick-walled EPS foam insulation
  • Adjustable nylon carrying straps for hand or shoulder carrying
  • Includes U-tek refrigerant
  • Capacity: 1.6 cu. ft.
Model 486
  • Washable, weather-resistant nylon shell with thick-walled EPS foam insulation
  • With Velcro™ closure and carrying strap
  • Duo-Temp locking
  • Includes U-tek refrigerant
  • Capacity: 1.6 cu. ft.
Model 640
  • Washable, weather resistant nylon shell with polyethylene foam insulation
  • Inner partition divides unit to separate specimens
  • Built-in lid compartment for U-tek refrigerant packs
  • Reusable
  • Printed with standard biohazard symbol
  • Zipper lock closure
  • Duo-Temp locking
  • Includes U-tek refrigerant
Specifications
Height (Metric)21.59 cm
Height (English)8.5 in.
Length (Metric)27.3 cm
Length (English)10.75 in.
Width (Metric)19.6 cm
Width (English)7.75 in.
Dimensions (L x W x H)10.75 x 7.75 x 8.5 in. (27.3 x 19.6 x 21.59 cm)
Closure TypeZipper Lock
IncludesU-tek refrigerant, built-in lid compartment, Duo-Temp locking
MaterialNylon
TypeMedical Transporter
Unit SizeEA