T332 Micrewtube™ Microcentrifuge Tubes
Actual product may vary
Simport Scientific
T332 Micrewtube™ Microcentrifuge Tubes
T332 Micrewtube™ Microcentrifuge Tubes are graduated tubes with markup area for easy identification.
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-046-1010.5 mL
Catalog number FIS22-046-101
Price (SGD)
-
Capacity (Metric):
0.5 mL
  • Washer seal secured in the top of the cap
  • Ensures a positive leakproof seal, time after time
  • Caps are supplied with attachment loops in order to prevent mix-up and contamination
  • Used for cryogenic work
  • Withstand temperatures from -196°C to +121°C
Specifications
Certifications/ComplianceRNase, DNase, pyrogen-free
Capacity (Metric)0.5 mL
MaterialPolypropylene
IncludesO-ring seal, screw cap, and attachment loop
Label TypeWhite Marking Spot
Packaging500/Cs.
SterilitySterile
GraduatedNo
Capacity (English)0.016 oz.
Bottom ShapeSkirted
Max. RCF20,000 x g
Product TypeMicrocentrifuge Tube
Unit SizeCase