4mL (13mm) Screw Thread Vial Kits
4mL (13mm) Screw Thread Vial Kits
Actual product may vary
Thermo Scientific
4mL (13mm) Screw Thread Vial Kits
Manufactured from high quality glass, these 4mL (13mm), 15x45mm vials feature 13-425 thread finish and fit a variety of autosamplers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMarking SpotColorSeptumPackaging Type
FIS22-045948YesAmberWhite Virgin PTFE, 0.01 in.Polybag
FIS03-375-3GNoClearRed PTFE/White SiliconeTray
Catalog number FIS22-045948
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Marking Spot:
Yes
Color:
Amber
Septum:
White Virgin PTFE, 0.01 in.
Packaging Type:
Polybag
Manufactured from high quality glass, these 4mL (13mm) 15 x 45mm vials feature a 13-425 thread finish. Available in Thermo Scientific™ National™ and Chromacol™ brands.
Feature superior 51A amber (Type 1, Class AB) or 33 expansion borosilicate clear (Type 1, Class A) glass construction.
  • Microsampling and High Recovery Vials allow maximum sample extraction without need for separate inserts
  • Graduated, write-on patch available for convenient sample identification
  • Optional silanized (deactivated) glass maximizes recovery of critical polar, labile or OH-interacting compounds
  • Polyspring inserts are self-aligning and provide a cushion against needle contact
Recommended for the following instruments:
  • Thermo Scientific™ Dionex™
  • Shimadzu™
  • Spark Holland™
  • Varian™
  • VWR (Merck)/Hitachi™
  • Waters™ (Wisp 48 Position Carousel)
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure ColorBlack
Diameter (Metric) Inner15 mm
ColorAmber
MaterialGlass, PTFE
Pre-slitNo
Volume (Metric)4 mL
SeptumWhite Virgin PTFE, 0.01 in.
IncludesVial: C4015-2W, Closure: C4015-1A/C4015-10
Packaging TypePolybag
Volume (Metric) Total4 mL
Closure MaterialPolypropylene
Closure TypeScrew Cap, Open Top, 8.5 mm hole, Patched
Height (Metric)45 mm
TypeScrew Top Vial Kit
Closure Size13-425
Marking SpotYes
Septa Size (Metric)13-425 mm
Unit SizePK