Autoclavable HD Biohazard Waste Bags
Autoclavable HD Biohazard Waste Bags
Actual product may vary
Autoclavable HD Biohazard Waste Bags
Strong and leak-resistant
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorDimensions (L x W)Thickness
FIS22-044565Red with Black Print36 x 45 in. (91 x 114 cm)1.50 mil
FIS22-044559Clear with Red Print30 x 24 in. (76 x 61 cm)1.50 mil
FIS22-04-4558Clear with Red Print24 x 12 in. (61 x 30 cm)1.50 mil
FIS22-044561Red with Black Print12 x 8 in. (30 x 20 cm)1.25 mil
FIS22-044560Clear with Red Print36 x 24 in. (91 x 61 cm)1.50 mil
FIS22-044563Red with Black Print23 x 19 in. (58 x 48 cm)1.25 mil
FIS22-044564Red with Black Print35 x 25 in. (89 x 64 cm)1.50 mil
Catalog number FIS22-044565
Price (SGD)
377.00
CS
Add to cart
Color:
Red with Black Print
Dimensions (L x W):
36 x 45 in. (91 x 114 cm)
Thickness:
1.50 mil
Price (SGD)
377.00
CS
Add to cart
Strong and leak-resistant
  • Printed with biohazard symbol and text in English and Spanish
  • Supplied with twist tie closure
Specifications
Closure TypeTwist Tie
ColorRed with Black Print
Dimensions (L x W)36 x 45 in. (91 x 114 cm)
Length (Metric)114 cm
Width (Metric)91 cm
AutoclavableAutoclavable
IncludesBiohazard symbol and text in English and Spanish
Width (English)45 in.
Length (English)36 in.
Thickness1.50 mil
MaterialPolyethylene
Unit SizeCS