Lithium Heparin Tubes
Actual product may vary
Lithium Heparin Tubes
Have Questions?
Catalog NumberCapacity (Metric)
FIS22-040-1043 mL
Catalog number FIS22-040-104
Price (SGD)
-
Capacity (Metric):
3 mL
Specifications
Product TypePlasma Lithium Heparin Blood Collection Tube
MaterialPET
Capacity (Metric)3 mL
Diameter (Metric) Inner13 mm
For Use With (Application)For Blood collection
Length (Metric)75 mm
Length (English)2.95 in.
Certifications/ComplianceCE
Unit SizePack