VACUETTE™ Heparin Blood Collection Tubes
Actual product may vary
VACUETTE™ Heparin Blood Collection Tubes
VACUETTE™ Heparin Blood Collection Tubes have a green top and contain spray-dried lithium, sodium or ammonium heparin on the inner walls and are usedin clinical chemistry, immunology and serology. The anticoagulant heparin activates antithrombin, which blocks the clotting cascade and thus produces a whole blood/plasma sample.
Have Questions?
Catalog NumberCapacity (Metric)Closure TypeAdditive
FIS22-040-0212 mLNon-ridged Pull CapLithium Heparin
Catalog number FIS22-040-021
Price (SGD)
-
Capacity (Metric):
2 mL
Closure Type:
Non-ridged Pull Cap
Additive:
Lithium Heparin
VACUETTE™ Heparin Blood Collection Tubes have a green top and contain spray-dried lithium, sodium or ammonium heparin on the inner walls and are usedin clinical chemistry, immunology and serology. The anticoagulant heparin activates antithrombin, which blocks the clotting cascade and thus produces a whole blood/plasma sample.
  • Tubes are supplied in packs of 50 pieces
  • For venus blood collection
  • Primarily used for drawing blood for chemistry and immunochemistry testing
  • Made out of virtually unbreakable PET plastic
  • Provides improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • VACUETTE™ Lithium Heparin Blood Collection Tubes are also available with a gel separator
  • Heparin tubes from Greiner Bio-One are available in: LH Lithium Heparin, LH Lithium Heparin Sep and NH Sodium Heparin.
  • CE marked
Specifications
Product TypeBlood Collection Tube
AdditiveLithium Heparin
MaterialPET
SterilitySterile
Closure TypeNon-ridged Pull Cap
Capacity (Metric)2 mL
For Use With (Equipment)VACUETTE™ Blood Collection Needles
Closure ColorGreen with Black Ring
Height (Metric)75 mm
Tube Size13 x 75 mm
For Use With (Application)Blood Collection
Diameter (Metric) Outer13 mm
Unit SizePack