Tissue Path™ Disposable Embedding Rings
Tissue Path™ Disposable Embedding Rings
Actual product may vary
Tissue Path™ Disposable Embedding Rings
Tissue Path™ Disposable Embedding Rings fit both shallow and deep base molds.
Have Questions?
Catalog NumberColor
FIS22-038197White
Catalog number FIS22-038197
Price (SGD)
254.00
CS
Add to cart
Color:
White
Price (SGD)
254.00
CS
Add to cart
  • High-impact polystyrene ensures accurate positioning of tissue samples
  • Etched marking surface can be easily written on for positive sample identification
  • Rings fit both shallow and deep base molds
Specifications
Product TypeEmbedding Ring
MaterialPolystyrene
ColorWhite
DisposableSingle-use
For Use With (Application)Designed for use with both shallow and deep base molds, for holding and identifying tissue sample blocks and fit well in microtome chuck adapters
Quantity250 Pk.
Unit SizeCS