Superfrost™ Plus Microscope Slides
Actual product may vary
Superfrost™ Plus Microscope Slides
Superfrost™ Plus Microscope Slides
Superfrost™ Plus Microscope Slides
Superfrost™ Plus Microscope Slides
Superfrost™ Plus Microscope Slides
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears.
Have Questions?
Catalog NumberCorner Style
FIS22-037-246Clipped
Catalog number FIS22-037-246
Price (SGD)
-
Corner Style:
Clipped
An ideal choice for frozen sections, cytocentrifuge preparations, and standard Papanicolaou smears
Same outstanding features as regular Superfrost slides with special treatment process that electrostatically adheres tissue sections and cells to the glass without the need for adhesives or protein coatings.
  • Treatment virtually eliminates tissue loss during staining; saves hours of slide preparation
  • Chemical-resistant white marking surface
  • Size: 25 × 75mm
Specifications
FrostedYes
Length (Metric)75 mm
MaterialGlass
TypeMicroscope Slide
Width (Metric)25 mm
ColorWhite
Corner StyleClipped
Quantity2 Packs of 72
Unit SizePack