Colormark™ Slide Cartridges forSHUR/Mark™ Labeling Systems
Colormark™ Slide Cartridges forSHUR/Mark™ Labeling Systems
Actual product may vary
Colormark™ Slide Cartridges forSHUR/Mark™ Labeling Systems
Background color makes etched patient information more visible
Have Questions?
Catalog NumberColorCoating
FIS22-035815BlueNon-adhesion
Catalog number FIS22-035815
Price (SGD)
158.00
PK
Add to cart
Color:
Blue
Coating:
Non-adhesion
Price (SGD)
158.00
PK
Add to cart
  • Brightly coated with contrasting background color
  • Self-contained packages ready for loading into your SHUR/Mark slide labeling system
  • Simply remove the cover from the slide package and drop the sleeve into the loading bin of the SHUR/Mark labeling system
  • Available with Plus formula to provide the utmost adhesion
Specifications
ColorBlue
CoatingNon-adhesion
Quantity144 Pk., 10 Pk./Cs.
Unit SizePK