Biohazard Disposal Bags with or without Warning Label
Biohazard Disposal Bags with or without Warning Label
Actual product may vary
Bel-Art
Biohazard Disposal Bags with or without Warning Label
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (L x W)ColorIncludes
FIS22-03292636 x 24 in. (91 x 61 cm)TranslucentNon-printed w/o Sterilization Patch
FIS13-717-28724 x 12 in. (61 x 30 cm)TranslucentNon-printed w/o Sterilization Patch
FIS13-717-28924 x 12 in. (61 x 30 cm)ClearBiohazard Symbol w/ Sterilization Patch
FIS13-717-29136 x 24 in. (91 x 61 cm)ClearBiohazard Symbol w/ Sterilization Patch
Catalog number FIS22-032926
Price (SGD)
636.00
PK
Add to cart
Dimensions (L x W):
36 x 24 in. (91 x 61 cm)
Color:
Translucent
Includes:
Non-printed w/o Sterilization Patch
Price (SGD)
636.00
PK
Add to cart
Economical, safe disposal of used pipettes, petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters, and other biohazard wastes
  • These transparent, polypropylene plastic biohazard bags are 0.03 mm (1.5 mil) thick
  • High temperature autoclavable at a maximum of 135°C (275°F)
  • May also be used as conventional trash can liners
Specifications
AutoclavableAutoclavable
Capacity (English)20 gal.
Capacity (Metric)170 L
Certifications/ComplianceASTM D1709-98
Closure TypeZip Tie
ColorTranslucent
DisposableSingle-use
For Use With (Application)Disposal of used pipettes, petri dishes, culture plates, membrane filters and more
IncludesNon-printed w/o Sterilization Patch
MaterialPolypropylene
SterilityNon-sterile
Thickness (Metric)1.5 mm
Thickness (English)1.5 mil
Dimensions (L x W)36 x 24 in. (91 x 61 cm)
Length (English)36 in.
Length (Metric)91 cm
Width (English)24 in.
Width (Metric)61 cm
Product TypeBiohazard Bag
Unit SizePK