LeLoop™ Holder
LeLoop™ Holder
Actual product may vary
LeLoop™ Holder
Holders for LeLoop Inoculating Loops
Have Questions?
Catalog NumberLength (English)
FIS22-0320958 in.
Catalog number FIS22-032095
Price (SGD)
-
Length (English):
8 in.
  • Nickel-plated brass with insulated handles
  • Available in 6 and 8 in. lengths
Specifications
Length (English)8 in.
Length (Metric)20.3 cm
MaterialNickel-plated Brass
SterilityNon-sterile
SizeLarge
Product TypeInoculating Loop Holder
For Use With (Equipment)LeLoop™ inoculating loop
Unit SizeEA