LeLoop™ Inoculating Loops
LeLoop™ Inoculating Loops
Actual product may vary
LeLoop™ Inoculating Loops
More economical than platinum or disposable plastic loops
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolumeCalibratedDiameter (Metric) Inside
FIS22-0320915.45 μLNo0.35 cm
FIS22-03208810 μLYes0.4 cm
Catalog number FIS22-032091
Price (SGD)
-
Volume:
5.45 μL
Calibrated:
No
Diameter (Metric) Inside:
0.35 cm
More economical than platinum or disposable plastic loops
Reusable loops are made of high-quality twisted nichrome wire.
  • Loops slide smoothly across agar surfaces without tearing or gouging
  • The nichrome wire cools rapidly, saving time
  • Loops are available calibrated or noncalibrated
  • Calibrated in accordance with ASM/APHA via Evans Blue Test and include calibration certificates
  • LeLoop™ loops come without holders, which are available separately
Specifications
Diameter (English) Inside0.14 in.
Diameter (Metric) Inside0.35 cm
Length (English)3 in.
Length (Metric)7.62 cm
MaterialNichrome
SterilityNon-sterile
Volume5.45 μL
SizeLarge
Product TypeInoculating Loop
CalibratedNo
Unit SizePK